ZING Green Safety Eco Safety Sign, Fire Hose w/Arrow
Supplier: ZING Green Safety Products
2893S 1893 1893G 1893S 1893A 2893 2893G 2893A
76023-188EA 24.78 USD
76023-188 76025-606 76023-194 76025-198 76023-190 76025-200 76023-192 76025-202
ZING Green Safety Eco Safety Sign, Fire Hose w/Arrow
Signs
The sign is available in two sizes, 10"x7" and 14"x10", and four eco-friendly materials.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Glow-in-the-Dark signs are manufactured from recycled plastic with a high-performance, safety grade photoluminescent coating. Signs will glow for a period of 10 hours on a full charge and require no electrical power source.
The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
Certifications: OSHA 1910.158(c)(1)