SHOWA 747 Nitrile, Chemical Resistant, 22 mil, 18" Glove, Showa
Supplier: Showa
747-11
89041-905CS 1540.06 USD
89041-905
SHOWA 747 Nitrile, Chemical Resistant, 22 mil, 18" Glove, Showa
Gloves
100% Nitrile gloves provide excellent protection from chemicals, cuts, punctures, and snags.
  • 100% Nitrile
  • 18" length
  • Food safe
These gloves are designed for handling most solvent-based chemicals, in a wide range of industries.