SHOWA Atlas 660 Fully Coated, Triple Dipped, PVC, Chemical Resistant Glove, 12", Showa
Supplier: Showa
660S-07 660M-08 660L-09 660XL-10 660XXL-11
10847-752CS 571.77 USD
10847-752 10847-754 10847-756 10847-758 10847-760
SHOWA Atlas 660 Fully Coated, Triple Dipped, PVC, Chemical Resistant Glove, 12", Showa
Gloves
Excellent protection from liquids and chemicals used in manufacturing, finishing, maintenance, refining, and masonry.
  • Triple dipped PVC
  • 12" length
  • Fully coated
  • Extremely flexible at low temperatures
Extremely flexible proprietary PVC coating. Ideally suited for use where protection from liquids, chemicals, and abrasion is needed.