Hydrochloric acid 0,5% in 70% alcohol

Supplier: Electron Microscopy Sciences

Synonyms: Hydrochloric acid solution, Muriatic acid

26071-01 26071-04
100504-454EA 74.89 USD
100504-454 100504-882
Hydrochloric acid 0,5% in 70% alcohol
Hydrochloric acid

An acidified alcohol solution for the differentiation of regressive stains.

Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR