SHOWA 430 A4 Cut Resistant Glove, Nitrile Palm Coated, Showa
Supplier: Showa
430-07 430-08
75856-504CS 1757.16 USD
75856-504 75856-506 75856-508 75856-510 75856-514
SHOWA 430 A4 Cut Resistant Glove, Nitrile Palm Coated, Showa
Gloves
Ideal glove for handling sharp materials, such as metal and glass.
  • Excellent dexterity
  • Foam Nitrile coating
  • ANSI A4 cut resistance
Excellent glove for protection from oils, hydrocarbons, grease, and abrasion. Use when handling sharp edges and dexterity is required.
Certifications: ANSI Cut Level A4.