Solvent Blue 38 0.1% in 95% alcohol with acetic acid

Supplier: Electron Microscopy Sciences

Synonyms: Direct Blue 86Luxol® Fast Blue MBSNC.I. 74180

26056-10
101410-934EA 75.21 USD
101410-934
Solvent Blue 38 0.1% in 95% alcohol with acetic acid
Solvent Blue 38
Formula: C₃₂H₁₂CuN₈Na₂O₆S₂
MW: 778.15 g/mol
MDL Number: MFCD00071424
CAS Number: 1328-51-4
https://us.vwr.com/stibo/web/std.lang.all/48/46/28984846.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR