Collodion 2% in amyl acetate, Ultrapure, sterile
Supplier: Electron Microscopy Sciences
12620-10
12620-55
12620-50
100496-438EA
126.45
USD
100496-438
100496-446
100496-444
Collodion 2% in amyl acetate, Ultrapure, sterile
Collodion
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail. Two types are available: Our sterile formula which is filtered down to 0.45 micron and our non-sterile formula.
Ordering information: Collodion (Parlodian) offered in a 30 ml bottle to avoid its classification as a hazardous chemical for shipping purposes.
MDL Number:
MFCD00081525 CAS Number: 9004-70-0 UN: 2059 ADR: 3,II |