Basic fuchsin 0.1% in methanol 95%

Supplier: Electron Microscopy Sciences

Synonyms: Basic-fuchsinPararosanilineFuchsin basicBasic fuchsin hydrochlorideDiamondfuchsinBasic Red 9(4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochlorideBasic fuchsinCI 425104-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochlorideMagenta ORosaniline hydrochloride

26031-10
101410-892EA 86.84 USD
101410-892
Basic fuchsin 0.1% in methanol 95%
Basic Fuchsin

Basic Fuchsin solution Electron Microscopy Sciences is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology.
Available in concentrations of 0.25% aqueous, 0.5% aqueous, 0.1% in 95% methanol, and as a working solution.

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
MDL Number: MFCD00012569
CAS Number: 632-99-5
https://us.vwr.com/stibo/web/std.lang.all/47/83/28984783.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR