SHOWA BO500B Ergonomic Design, Polyurethane Coated Product Protection Gloves, Black, Showa
Supplier: Showa
BO500B-XL BO500B-S BO500B-L
89065-434CS 563.61 USD
89065-434 89065-428 89065-432
SHOWA BO500B Ergonomic Design, Polyurethane Coated Product Protection Gloves, Black, Showa
Gloves
Proprietary ergonomic design provides unequalled comfort and dexterity.
  • Polyurethane palm coated
  • 13 gauge Nylon liner
  • Ergonomic design
These gloves are used in clean environments. They are lightweight for delicate part handling and inspection, with a polyurethane coating that provides excellent dry/oil grip.
Packaging: Individually poly-bagged.