N-(2-Diethylaminoethyl)-1-naphthylamine oxalate

Supplier: Pfaltz & Bauer

Synonyms: Tsuda's ReagentN,N-Diethyl-N'-1-naphthylethylenediamine OxalateN1,N1-Diethyl-N2-(naphthalen-1-yl)ethane-1,2-diamine oxalate

D23832-5G
101182-108EA 1003.12 USD
101182-108
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
Formula: C₁₆H₂₄N₂O₄
MW: 332.40 g/mol
MDL Number: MFCD00060156
CAS Number: 29473-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR