Casein Hydrolysate Acid Solution, Vitamin Free, MP Biomedicals
Supplier: MP Biomedicals
0210476990
75875-610EA 216.26 USD
75875-610
Casein Hydrolysate Acid Solution, Vitamin Free, MP Biomedicals
Media for Microbiology Additives and Supplements for Microbiology
This casein hydrolysate is an hydrochloric acid digest of casein in which the protein molecule has been substantially reduced to its constituent amino acids.
Laboratory media including basal media for vitamin analysis of folic acid, pantothenic acid, riboflavin, thiamin, niacin, and B-12.
Store at room temperature. Clear solution