Hematoxylin in 95% alcohol

Supplier: Electron Microscopy Sciences
Danger

Synonyms: (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaolHydroxybrasilinNSC 2785DcH-48Hydroxybrazilin(+)-HematoxylinNatural Black I

26042-10
101410-912EA 181.5 USD
101410-912
Hematoxylin in 95% alcohol
Hematoxylin

Hematoxylin solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the biological staining commission and the armed forces institute of pathology.


  • Available in concentrations of 1.0% aqueous, 5% in SD alcohol, in 95% alcohol, and as Heidenhain's hematoxylin

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2
UN: 1170
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/48/30/28984830.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR