Difco™ Sabouraud Brain Heart Infusion Agar Base, BD
Supplier: BD Diagnostics
279720 279720
90004-018EA 488.85 USD
CA90004-018 90004-018
Difco™ Sabouraud Brain Heart Infusion Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Use with chloromycetin and blood (optional) in cultivation and isolation of pathogenic fungi
It can also used in qualitative procedures for cultivation of dermatophytes and other pathogenic and non-pathogenic fungi from clinical and non-clinical specimens.