HEPES sodium salt ≥99.0% for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219482825 0219482880 0219482890
IC0219482825EA 87.65 USD
IC0219482825 IC0219482880 IC0219482890
HEPES sodium salt ≥99.0% for molecular biology
Formula: C₈H₁₇N₂NaO₄S
MDL Number: MFCD00036463
CAS Number: 75277-39-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25fedeb8d015d893ba4bc84e85015b582293fdd8&vid=1213b6ccdaee8ce22d032b647e5d07e2d376fe69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Physical Appearance White Powder
pH 9.5 - 10.8 (1% aq soln); Useful pH range=6.8-8.2(Lit)
pKa 7.55 at 25 °C (Lit.)
Loss on Drying <3.0%
Solubility Soluble in water.
Sterilization of Solutions Solutions may be autoclaved under standard conditions


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR