Silver nitrate 0.01 N aqueous solution

Supplier: AQUA SOLUTIONS MS

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

8387-4L 8387-500ML 8387-1L 8387-20L 8387-10L
101226-528EA 88.89 USD
101226-528 101226-156 101226-418 101225-812 101225-712
Silver nitrate 0.01 N aqueous solution
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Prepared to formulation on file Yes
Actual Normality 0.0097 - 0.0103
N.I.S.T. Traceable to SRM 919 Ye
Actual Density 0.99946 - 0.99976
Appearance and color Clear water white liquid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR