Odyssey® Pen, LI-COR®
Supplier: LI-COR®
926-71804
102673-444EA 35.69 USD
102673-444
Odyssey® Pen, LI-COR®
Pens Autoradiography Pens
Pen is suitable for writing on membranes to be scanned on any of the Odyssey Imagers or on Aerius infrared imaging systems.