Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
|
Synonyms:
Polyoxymethylene
Packaged under nitrogen.
0219998380
0219998320
IC0219998380EA
494.45
USD
IC0219998380
IC0219998320
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Paraformaldehyde
Methanol free Paraformaldehyde is ready to use solution used as a fixative for electron microscopy.
Recommended for use in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies.
Formula:
(CH₂O)n Storage Temperature: Refrigerator |
MDL Number:
MFCD00133991 CAS Number: 30525-89-4 UN: 2213 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...