SpectroQuest™ UV/VIS Scanning Spectrophotometer
Supplier: United Product & Instrument
SQ2800 SQ2800-515 SQ2800-528 SQ2800-401
470227-662EA 9495.35 USD
470227-662 470227-674 470227-676 470227-678
SpectroQuest™ UV/VIS Scanning Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The SpectroQuest™ 2800 is the most economic general-purpose UV/VIS spectrophotometer on the market. It is a stand-alone model with 4 nm fixed bandwidth and provides excellent performance for measurements in the range of 190 nm to 1100 nm. It has a large angled LCD screen with contrast adjustment for comfortable viewing in a variety of room conditions. The large sample compartment (100 mm) accommodates a wide range of cell holders and accessories including peristaltic sipper and peltier system. Optional PC download software and PC Windows® application software make this instrument veryMore Product Information