Micro Tubes, SafeSeal, SARSTEDT
Supplier: Sarstedt
72.695.001 72.695.002 72.695.007 72.695.004 72.695.006 72.695.005
101093-270PK 75.86 USD
101093-270 101093-272 101093-274 101093-276 101093-266 101093-268
Micro Tubes, SafeSeal, SARSTEDT
Tubes Microtubes
The safe seal locking cap mechanism ensures a secure seal for routine use and ideally suited for thermal applications.
  • Wide hinge for easy and precise closure
  • Large writing space on lid, graduated
  • Centrifugation to 30000 g (20 ml tubes: to 25000 g)