Anti-DiBrY Monoclonal Antibody [clone: 3A5]

Supplier: AdipoGen Life Sciences
JAI-MBY-020P
102979-286EA 344.77 USD
102979-286
Anti-DiBrY Monoclonal Antibody [clone: 3A5]
Antibodies
Neutrophils and eosinophils play an important role in the defence system against microbial infection. Myeloperoxidase (MPO) and eosinophil peroxidase (EPO) are known to catalyze formation of hypochlorous acid (HOCl) and hypobromous acid (HOBr). These reactive intermediates may react with proteins, lipids and nucleotides, and it has been reported to form tyrosine halogenation such as dibromotyrosine (DiBrY). DiBrY is a Br-modified tyrosine at 3- and 5- position, which is one of the major oxidative products derived from neutrophil myeloperoxidase. DiBrY may be a novel biomarker for tissue damage which is related to inflammatory and allergic disorders.

Type: Primary
Antigen: DiBrY
Clonality: Monoclonal
Clone: 3A5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: All, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR