Anti-CD56 Monoclonal Antibody (R-PE) [clone: ANC7C7]

Supplier: AdipoGen Life Sciences
ANC-308-050
102512-914EA 516.46 USD
102512-914
Anti-CD56 Monoclonal Antibody (R-PE) [clone: ANC7C7]
Antibodies
Human CD56 is an adhesion molecule from the Ig superfamily which is restricted to NK cells and a subset of T cells in the immune system. It is believed that NK cells form a first line of defense against tumor cells and cells infected with bacteria and viruses.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: ANC7C7
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR