Anti-CD66e Monoclonal Antibody (Biotin) [clone: CB30]

Supplier: AdipoGen Life Sciences
ANC-218-030
102512-540EA 346.15 USD
102512-540
Anti-CD66e Monoclonal Antibody (Biotin) [clone: CB30]
Antibodies
Human CD66e is a heavily glycosylated GPI anchored protein capable of heterophilic and homophilic adhesion and may play a role in metastasis of cancer cells. Soluble CD66e is also known as carcinoembryonic antigen (CEA).

Type: Primary
Antigen: CD66e
Clonality: Monoclonal
Clone: CB30
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR