Reinecke's salt ≥93.0%, dark red powder ACS

Supplier: CHEM IMPEX INTL INC MS

22922-100G 22922-25G
102563-840EA 224.83 USD
102563-840 102598-994
Reinecke's salt ≥93.0%, dark red powder ACS
Reinecke's salt

Reinecke salt can be used as precipitant for primary and secondary amines, proline, hydroxyproline, and certain amino acids.


Synonyms: Ammonium tetrarhodanatodiamminechromate(III) monohydrate; Ammonium tetrathiocyanatodiamminechromate(III) monohydrate.

Formula: C₄H₁₀CrN₇S₄
Storage Temperature: Ambient
MDL Number: MFCD00151654
CAS Number: 13573-16-5

Order Now

Specification Test Results

Appearance Dark red powder
Solubility Soluble in hot water; sparingly soluble in cold water
Sensitivity To pass test
Insoluble material ≤ 0.05% in dilute HCl
Purity limit ≥93.0% (Titration)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR