(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine ≥97%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 3-Aminopyrrolidine-1-carboxylic acid benzyl ester, Benzyl-3-aminopyrrolidine-1-carboxylate, 1-Cbz-3-aminopyrrolidine, 1-Z-3-aminopyrrolidine, 1-Benzyloxycarbonyl-3-aminopyrrolidine

17115-250MG 17115-1G 17115-25G 17115-5G
102567-678EA 64.12 USD
102567-678 102567-680 102612-416 102642-136
(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine ≥97%
(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

Synonym: 3-Aminopyrrolidine-1-carboxylic acid benzyl ester

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD03001673
CAS Number: 185057-50-5

Order Now

Specification Test Results

Appearance Yellow liquid
Purity limit ≥97%
Storage Temperature Store at 0 - 8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR