Brady® J50 Series Printer Cartridge
Supplier: Brady Worldwide
148763 148762 148764 148766
75846-018EA 83.59 USD
75846-018 75846-020 75846-024 75846-204
Brady® J50 Series Printer Cartridge
Inks
Large J50 series ink cartridges reduce the amount of changeovers or replacements needed.
  • Combination of the four CMYK ink cartridges produces 16 million color possibilities
  • Ink volume can allow for the printing of thousands of labels before the ink runs out depending on size and ink coverage of the label
  • Four separate ink cartridges allow the usage of all of the ink in each cartridge and individual replacement as needed
  • Ink cartridge replacement is simple and efficient, and can be completed in less than 10 seconds
Each color cartridge is separate toMore Product Information