Brady® BMP®61 Series Printer Ribbon R4400 Resin
Supplier: Brady
M61-R4410-WT 148132 148131 M61-R4410-BL 148133 M61-R4410-RD
76520-190RL 61.35 USD
76520-190 75840-071 75840-069 76520-192 76520-194 75840-067
Brady® BMP®61 Series Printer Ribbon R4400 Resin
Printer Ribbons
BMP®61 Series ribbon cartridges allow users to change ribbons in seconds.
  • One-color ribbon for printing a single color onto a label
  • R4400 Series Resin Formulation offers excellent resistance to solvents, scratches, and high heat when printed on the appropriate label material
  • Automatic label formatting with smart cell technology and easy loading
Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady®.