Yeast Malt Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M425-500G M425-2.5KG M425-5KG
95021-806EA 182.68 USD
95021-806 89225-482 89225-484
Yeast Malt Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Yeast malt broth (YM broth) is used for the isolation and cultivation of yeasts, moulds and other aciduric microorganisms.