SP Wilmad-LabGlass PEEK Union, SP Industries
Supplier: SP Industries
BP-1824-018
14007-326EA 53.46 USD
14007-326
SP Wilmad-LabGlass PEEK Union, SP Industries
Fittings and Connectors
This PEEK (polyether ether ketone) union is used for connecting 1/4"-28 fittings.
Ordering information: Nuts and ferrules included.