Benzalkonium chloride ~97%, waxy solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

0219015880 0219015883 0219015891
IC19015880EA 77.13 USD
IC19015880 IC19015883 IC19015891
Benzalkonium chloride ~97%, waxy solid
Benzalkonium chloride

Benzalkonium chloride is a cationic surface-active agent and germicide.


It is not effective against gram-negative bacteria or picornaviruses and it is not recommended for treatment of sanitizing critical or semicritical medical instruments.It has been reported that benzalkonium chloride inactivates HIV and HSV in vitro.Incompatible with anionic detergents, such as soap, and with nitrates. The preservative effect can be neutralized by the addition of Tween 80 and lecithin. The mode of action of quaternary ammonium compounds appears to be associated with the agent's effect on the cytoplasmic membrane, which controls cell permeability.


Soluble in water, alcohol, acetone; slightly soluble in benzene.


Caution: Research Use Only (RUO).

Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Boiling Pt: >100 °C/760 mmHg
Melting Pt: 34…37 °C (lit.)
Density: 0.98
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 2923
ADR: 8 III

Order Now

Specification Test Results

Identity Test Passes
Purity ~97%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR