β-Cyclodextrine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

0219005325 0219005380
IC19005325EA 101.39 USD
IC19005325 IC19005380
β-Cyclodextrine, white powder
β-Cyclodextrine

β-cyclodextrin is most commonly used as a complexing agent. It is reported to be useful for the selective precipitation of enantiomeric, positional or structural isomers. It also forms a water soluble complex with dansyl chloride for fluorescent labeling of proteins.


Cyclodextrins are cyclic oligosaccharides consisting of 6, 7, or 8 glucopyranose units with hydrophobic interiors, usually referred to as α-, β-, or γ-cyclodextrins, respectively. Lipophilic drugs of a size compatible with the hydrophobic core of a cyclodextrin can form complexes, resulting in increased aqueous solubility of the drugs. Cyclodextrins are non-toxic in many species (mice and rabbits), and do not denature proteins or interfere with enzymatic reactions.


Soluble in water, 1 M NH4OH: 50 mg/mL (Clear,Colorless Solution).

Formula: C₄₂H₇₀O₃₅
Melting Pt: 298…300 °C (lit.)
Density: 1,624 g/cm³ (lit.)
Flash Pt: 885.873 °C (Lit.)
Storage Temperature: Ambient
MDL Number: MFCD00078139
CAS Number: 7585-39-9
EINECS: 231-493-2

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 5 - 7.5
Loss on Drying ≤14%
Optical Rotation 158 ± 6° (c=1,water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR