Screw Thread Vials, Chemglass
Supplier: Chemglass
CV-2600-1232 CV-2590-1232 CV-2605-1232 CV-2595-1232 CV-1116-1232 CV-1117-1232 CV-1119-1232 CV-1120-1232
10170-622PK 90.38 USD
10170-622 10170-624 10171-022 10171-026 75820-392 75820-394 75820-396 75820-398
Screw Thread Vials, Chemglass
Vials
Standard opening screw thread vials contain a narrow opening allowing the use of GPI 8-425 closures and septa.
  • Available with graduated marking spots
  • Vials are available in silanized form
This vial is designed to work in a variety of auto samplers requiring narrow neck vials.