Cold Traps, PerkinElmer
Supplier: PerkinElmer
N9306093 L4271205 L4275107 N6101474 L4275110 N6200150 M0413535 L4275108 L4270123 N9306095 N6301054 L4271106 M0413628 L4275009 L4071151 M0413627 L4271290 N9306096 L4271203 N9306094 L1003028 L4275072 L1003027 L4271187
97019-164EA 1218.47 USD
97019-164 97019-160 97013-846 97013-638 97013-156 97013-144 97013-012 97013-034 97013-840 97013-036 97013-712 97019-628 97019-306 97012-816 97019-626 97012-814 97019-624 97013-140 97019-322 97013-154 97019-146 97019-630 97012-818 97019-158
Cold Traps, PerkinElmer
Vacuum Traps
TurboMatrix thermal desorption trap supplies exceptional analytical performance, used for U.S.
EPA Method TO17, the PerkinElmer standard trap, packed with Tenax™, on the TurboMatrix™ thermal desorber, will improve productivity and trapping capacity. The TurboMatrix air monitoring trap is packed with carbonaceous sorbents suitable for ozone precursor and air toxics monitoring.