Uvex Mercury™ Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S1502 S1502X
10790-524PK 62.52 USD
10790-524 10790-528 10790-530 10790-532 10790-556 10790-632 10790-634 10790-636
Uvex Mercury™ Safety Eyewear, Honeywell Safety
Safety Glasses
Uvex Mercury™ safety eyewear provides a comfortable fit and modern styling with clean lines design, substantial coverage and lens tint options.
  • Wraparound dual lens design offers panoramic field of vision and added side protection
  • Anti-scratch hardcoat for added lens life and durability or Uvextra®
  • Anti-fog coating for clearer lenses in environments where lens fogging occurs
  • Nosepiece with soft flexible 'fingers' adjusts
Certifications: ANSI Z87.1-2010 - meets the ANSI Z87.1-2010 standard and CSA Z94.3 - certified to the requirements of the CSA Z94.3 standard.