Middlebrook 7H10 Agar Base, HiMedia
Supplier: HiMedia
M196-500G M196-2.5KG M196-5KG
61000-044EA 187.33 USD
61000-044 89224-758 89224-760
Middlebrook 7H10 Agar Base, HiMedia
Media for Microbiology Prepared Media for Microbiology
Middlebrook 7H10 agar base used for isolation, cultivation and sensitivity testing of Mycobacterium tuberculosis.
  • Light yellow to light green homogeneous free flowing powder
  • Firm, comparable with 1.5% Agar gel