1-Methylindole ≥98%, light yellow, clear liquid
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
1-Methyl-1H-indole
, N-Methylindole
22008-1KG
22008-100G
22008-250G
22008-25G
102580-972EA
181.37
USD
102580-972
102568-650
102609-160
102580-970
1-Methylindole ≥98%, light yellow, clear liquid
1-Methylindole
Formula:
C₉H₉N MW: 131.18 g/mol Boiling Pt: 236…239 °C Density: 1.05 Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005800 CAS Number: 603-76-9 UN: 3334 ADR: 9, |
Specification Test Results
Purity by HPLC | ≥98% |
Specific Gravity | 1.04-1.05 g/mL |
Appearance | Light yellow liquid |
Refractive Index | 1.606 - 1.608 |
Storage | Store at 0-8 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...