4-(4-Methoxyphenyl)-1(2H)-phthalazinone ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

25040250MG
100265-588EA 41.85 USD
100265-588
4-(4-Methoxyphenyl)-1(2H)-phthalazinone ≥98%, crystalline powder
4-(4-Methoxyphenyl)-1(2H)-phthalazinone

White crystalline powder


Store in a cool, dry place

Formula: C₁₅H₁₂N₂O₂
MW: 252.27 g/mol
Melting Pt: 244 °C
MDL Number: MFCD02660689
CAS Number: 57353-93-2
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR