n-Pentatriacontane ≥96%

Supplier: INDOFINE CHEMICAL COMPANY MS

05350025MG
100282-192EA 402.77 USD
100282-192
n-Pentatriacontane ≥96%
n-Pentatriacontane

Store in a cool, dry place

Formula: H₃C(CH₂)₃₃CH₃
MW: 492.96 g/mol
MDL Number: MFCD00059397
CAS Number: 630-07-9
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR