N-Myristoyl-D-sphingosine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: C14 Ceramide

5611215MG
100291-388EA 359.62 USD
100291-388
N-Myristoyl-D-sphingosine ≥98%
N-Myristoyl-D-sphingosine

Store in a cool, dry place

Formula: C₃₂H₆₃NO₃
MW: 509.90 g/mol
MDL Number: MFCD01320386
CAS Number: 34227-72-0
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR