Mixture Fo 7 ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

901107100MG
100275-776EA 418.46 USD
100275-776
Mixture Fo 7 ≥99%
Mixture Fo 7

Store in a cool, dry place

MDL Number: MFCD06202097
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR