Methyl lignocerate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Tetracosanoic Acid Methyl EsterMethyl TetracosanoateLignoceric Acid Methyl EsterMethyl tetracosanoate

202400100MG
100299-080EA 341.31 USD
100299-080
Methyl lignocerate ≥99%
Methyl lignocerate

Store in a cool, dry place

Formula: C₂₅H₅₀O₂
MW: 382.67 g/mol
MDL Number: MFCD00009351
CAS Number: 2442-49-1
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR