Methyl-13-methylpentadecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21151325MG
100274-932EA 562.31 USD
100274-932
Methyl-13-methylpentadecanoate ≥98%
Methyl-13-methylpentadecanoate

Store in a cool, dry place

Formula: C₁₇H₃₄O₂
MDL Number: MFCD00053775
CAS Number: 5487-50-3
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR