Lyso-sulfatide (bovine)NH4-salt ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

5611421MG
100291-398EA 2253.15 USD
100291-398
Lyso-sulfatide (bovine)NH4-salt ≥99%
Lyso-sulfatide (bovine)NH4-salt

Store in a cool, dry place

MW: 542 g/mol
MDL Number: MFCD02265394
CAS Number: 38621-58-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR