1-Dodecylpyridinium chloride ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Laurylpyridinium chlorideDodecylpyridinium chloride

38010025MG
100290-802EA 300.77 USD
100290-802
1-Dodecylpyridinium chloride ≥98%
1-Dodecylpyridinium chloride

Store in a cool, dry place

Formula: C₁₇H₃₀ClN
MW: 283.88 g/mol
MDL Number: MFCD00011987
CAS Number: 104-74-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR