Dibehenin ≥99%, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

322200100MG
100290-396EA 396.23 USD
100290-396
Dibehenin ≥99%, solid
Dibehenin

Solid


Store in Freezer

MDL Number: MFCD00673515
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR