7-Bromo-2H-1,4-benzothiazin-3(4H)-one ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-Bromo-2H-benzo[b][1,4]thiazin-3(4H)-one

250111GM
100269-522EA 47.08 USD
100269-522
7-Bromo-2H-1,4-benzothiazin-3(4H)-one ≥98%, crystalline solid
7-Bromo-2H-1,4-benzothiazin-3(4H)-one

Off white crystalline solid


Store in a cool, dry place

Formula: C₈H₆BrNOS
MW: 244.11 g/mol
Melting Pt: 204…206 °C
MDL Number: MFCD02660583
CAS Number: 90814-91-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR