7-Allyloxycoumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19295100MG
100267-050EA 32.69 USD
100267-050
7-Allyloxycoumarin ≥98%, powder
7-Allyloxycoumarin

Light rose powder


Store in a cool, dry place

Formula: C₁₂H₁₀O₃
Melting Pt: 83.7…84.8 °C
MDL Number: MFCD00449867
CAS Number: 31005-03-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR