6-(4-Chlorophenyl)pyridazin-3(2H)-one ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

25094250MG
100265-654EA 41.85 USD
100265-654
6-(4-Chlorophenyl)pyridazin-3(2H)-one ≥98%, crystalline solid
6-(4-Chlorophenyl)pyridazin-3(2H)-one

White crystalline solid


Store in a cool, dry place

Formula: C₁₀H₇ClN₂O
Melting Pt: 267…268.5 °C
MDL Number: MFCD02676708
CAS Number: 2166-13-4
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR