15-Hydroxypentadecanoic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

14151525MG
100284-208EA 602.85 USD
100284-208
15-Hydroxypentadecanoic acid
15-Hydroxypentadecanoic acid

Store in a cool, dry place

Formula: C₁₅H₃₀O₃
MW: 258.40 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00039535
CAS Number: 4617-33-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR