1,3-Dicaprylin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1,3-Dioctanoylglycerol(2-hydroxy-3-octanoyloxypropyl) octanoate

32080350MG
100290-248EA 381.85 USD
100290-248
1,3-Dicaprylin ≥99%
1,3-Dicaprylin

Store in a cool, dry place

Formula: C₁₉H₃₆O₅
MW: 344.492 g/mol
MDL Number: MFCD00056316
CAS Number: 1429-66-9
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR