Starch solution R Reag. Ph. Eur. 1085103
Supplier: Ricca Chemical
R8046000-500A
RC8046000-500AEA
165.8
USD
RC8046000-500A
Starch solution R Reag. Ph. Eur. 1085103
Starch
Used for oxidation-reduction analysis.
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol |
MDL Number:
MFCD00082026 CAS Number: 9005-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...